Please login to your online account to display your discounted pricing

1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil, ACROS Organics™


Quantity 10mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  361310100

 View more versions of this product

Catalog No. AC361310100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 260.37
Quantity 10mL
Color Clear
EINECS Number 221-111-2
TSCA TSCA
Assay 49 to 51%
Density 0.9500g/mL
Beilstein 07, III, 30
CAS Min % 49.0
CAS Max % 51.0
CAS Min % 49.0
CAS Max % 51.0
Health Hazard 2 GHS H Statement:
Heating may cause a fire.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep only in original container.
Wear protective gloves/protective clothing/eye protection/face protection.
Protect from sunlight.
Store away from other materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Chemical Name or Material 1, 1-Di(tert-butylperoxy)cyclohexane
Name Note 50% Solution in Mineral Oil
MDL Number MFCD00084800
CAS 3006-86-8
Health Hazard 1 GHS Signal Word: Danger
Additional Information hydroperoxides (as TBHP): < 0.2%
CAS 8042-47-5
Molecular Formula C14H28O4

Specifications

Chemical Name or Material 1, 1-Di(tert-butylperoxy)cyclohexane
Name Note 50% Solution in Mineral Oil
CAS 3006-86-8
CAS Min % 49.0
CAS Max % 51.0
CAS 8042-47-5
CAS Min % 49.0
CAS Max % 51.0
Assay 49 to 51%
Molecular Formula C14H28O4
Formula Weight 260.37
MDL Number MFCD00084800
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep only in original container.
Wear protective gloves/protective clothing/eye protection/face protection.
Protect from sunlight.
Store away from other materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Heating may cause a fire.

SDS & Safety Documents