missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 97%

Manufacturer:  Alfa Aesar™ H55957ME

 View more versions of this product

Catalog No. AAH55957ME



Description

Description

Applications
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons in vitro and in vivo because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

Solubility
Soluble in Dimethylformamide (DMF).

Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
500mg
C59H98ClN2O4+
8251870
Soluble in Dimethylformamide (DMF).
CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O
934.893
933.88
68°C (decomposition)
41085-99-8
MFCD00142354
1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
JVXZRNYCRFIEGV-UHFFFAOYSA-N
(2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid
129893813
97%

Chemical Identifiers

41085-99-8
934.893
JVXZRNYCRFIEGV-UHFFFAOYSA-N
129893813
CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O
C59H98ClN2O4+
MFCD00142354
1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
(2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents