Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Cyclopropanedicarboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 1,1-Cyclopropanedicarboxylic acid
CAS 598-10-7
Molecular Formula C5H6O4
Formula Weight 130.1
MDL Number MFCD00013727
UN Number UN3261
Beilstein 1864823
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1708506

  SDS
alfa aesar™
A1708506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.1
Chemical Name or Material 1,1-Cyclopropanedicarboxylic acid
Assay 97%
Molecular Formula C5H6O4
MDL Number MFCD00013727
CAS 598-10-7
UN Number UN3261
Beilstein 1864823
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 128° to 135°C
Quantity 5g

Applications
1,1-Cyclopropanedicarboxylic acid, is used as an inhibitor of 1-aminocyclopropane-1-carboxylic acid oxidase, was quantitated in Lycopersicum esculentum by HPLC-electrospray tandem mass spectrometry. Crystal and molecular structure of cyclopropane-1,1-dicarboxylic acid has been reported. Cyclopropane-1,1-dicarboxylic acid was used in the preparation of new heterocyclic derivatives of cyclopropane dicarboxylic acid containing thiadiazole and 1,2,4-triazole moieties. It was also used to prepare spiro-cyclopropyl metallocycles. It can also be used as a building block for the synthesis of a series of cyclopropanecarboxamides, having antifungal activity.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.