Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Cyclohexanedicarboxylic acid, 95%

$139.62 - $139.62

Chemical Identifiers

Chemical Name or Material 1,1-Cyclohexanedicarboxylic acid
CAS 1127-08-8
Molecular Formula C8H12O4
Formula Weight 172.18
MDL Number MFCD00094668
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2602503

 
alfa aesar™
H2602503
1g Each for $139.62
Description & Specifications

Specifications

Formula Weight 172.18
Chemical Name or Material 1,1-Cyclohexanedicarboxylic acid
Assay 95%
Molecular Formula C8H12O4
MDL Number MFCD00094668
CAS 1127-08-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Odorless
Melting Point ≈175°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.