missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Cyclohexanediacetic acid, 99%, ACROS Organics™

$929.83 - $929.83

Chemical Identifiers

CAS 4355-11-7
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00001519
InChI Key YQPCHPBGAALCRT-UHFFFAOYSA-N
Synonym 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
PubChem CID 78061
IUPAC Name 2-[1-(carboxymethyl)cyclohexyl]acetic acid
SMILES C1CCC(CC1)(CC(=O)O)CC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC173930250 SDS View Documents Acros Organics
173930250
25g Glass bottle Each for $929.83
Specifications

Chemical Identifiers

4355-11-7
200.23
YQPCHPBGAALCRT-UHFFFAOYSA-N
78061
C1CCC(CC1)(CC(=O)O)CC(=O)O
C10H16O4
MFCD00001519
1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
2-[1-(carboxymethyl)cyclohexyl]acetic acid

Specifications

Authentic
1% max.
181°C to 185°C
4355-11-7
100.0
C10H16O4
MFCD00001519
YQPCHPBGAALCRT-UHFFFAOYSA-N
2-[1-(carboxymethyl)cyclohexyl]acetic acid
78061
Crystalline Powder
1, 1-Cyclohexanediacetic acid, 99%
Glass bottle
White
25g
98.5
99%
C6H10(CH2CO2H)2
1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
C1CCC(CC1)(CC(=O)O)CC(=O)O
200.23
200.23
≥98.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 224-427-9

TSCA : TSCA