Please login to your online account to display your discounted pricing

1,1-Cyclobutanedicarboxylatodiammineplatinum(II), ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 1-Cyclobutanedicarboxylatodiammineplatinum(II)
CAS 41575-94-4
Molecular Formula C6H12N2O4Pt
Formula Weight 371.26
MDL Number MFCD00070464
Synonym Carboplatin
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377982500

  SDS
thermo fisher scientific
AC377982500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC377980010

  SDS
thermo fisher scientific
AC377980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline
Formula Weight 371.26
Quantity 250mg
Color Off-white
Merck Index 15, 1823
Molecular Formula C6H12N2O4Pt
MDL Number MFCD00070464
Synonym Carboplatin
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause genetic defects.
Suspected of causing cancer.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage the
Infrared Spectrum Authentic
Chemical Name or Material 1, 1-Cyclobutanedicarboxylatodiammineplatinum(II)
Platinum (Pt) 51.8% min.
CAS 41575-94-4
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause genetic defects.
Suspected of causing cancer.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage the