Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Carbonyldipyrrolidine, 98%

Chemical Identifiers

Chemical Name or Material 1,1'-Carbonyldipyrrolidine
CAS 81759-25-3
Formula Weight 168.24
MDL Number MFCD00216641
Beilstein 1637732
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6094209

 
alfa aesar™
H6094209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6094218

 
alfa aesar™
H6094218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.24
Boiling Point 148° to 150°C
Chemical Name or Material 1,1'-Carbonyldipyrrolidine
Density 1.086g/mL
Physical Form Liquid
Assay 98%
MDL Number MFCD00216641
CAS 81759-25-3
Beilstein 1637732
Refractive Index 1.513
Health Hazard 1 Warning
Sensitivity Light Sensitive
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Color Colorless to pale yellow
Quantity 10g

Applications
Reactant in the ring-opening polymerization of six-and seven-membered cyclic pseudoureas.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C