Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Carbonyldipiperidine, 99%

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material 1,1'-Carbonyldipiperidine
CAS 5395-04-0
Molecular Formula C11H20N2O
Formula Weight 196.29
MDL Number MFCD00006482
Synonym Bis(pentamethylene)urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2717314

  SDS
alfa aesar™
H2717314
25g Each for $226.01
Description & Specifications

Specifications

Formula Weight 196.29
Boiling Point 296° to 298°C
Quantity 25g
Synonym Bis(pentamethylene)urea
Chemical Name or Material 1,1'-Carbonyldipiperidine
MDL Number MFCD00006482
Assay 99%
Health Hazard 1 Warning
Melting Point 43° to 46°C
CAS 5395-04-0
Molecular Formula C11H20N2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.