Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(1-Boc-4-piperidinyl)-4-iodopyrazole, tech. 90%

Chemical Identifiers

Chemical Name or Material 1-(1-Boc-4-piperidinyl)-4-iodopyrazole
CAS 877399-73-0
Purity Grade Technical Grade
Formula Weight 377.23
MDL Number MFCD12913927
Beilstein 14511019
Synonym 1-Boc-4-(4-iodo-1H-pyrazol-1-yl)piperidine; tert-Butyl 4-(4-iodo-1H-pyrazol-1-yl)piperidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6591803

 
alfa aesar™
H6591803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6591806

 
alfa aesar™
H6591806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65918MD

 
alfa aesar™
H65918MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 377.23
Chemical Name or Material 1-(1-Boc-4-piperidinyl)-4-iodopyrazole
Beilstein 14511019
Purity Grade Technical Grade
Physical Form Solid
CAS 877399-73-0
Synonym 1-Boc-4-(4-iodo-1H-pyrazol-1-yl)piperidine; tert-Butyl 4-(4-iodo-1H-pyrazol-1-yl)piperidine-1-carboxylate
MDL Number MFCD12913927
Quantity 1g
Assay 90%
Sensitivity Light Sensitive