Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(1-Boc-3-azetidinyl)pyrrolidine, 94%

$89.73 - $494.11

Chemical Identifiers

Chemical Name or Material 1-(1-Boc-3-azetidinyl)pyrrolidine
CAS 1019008-21-9
Molecular Formula C12H22N2O2
Formula Weight 226.32
MDL Number MFCD09860616
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2711103

 
alfa aesar™
H2711103
1g Each for $494.11

AAH27111MD

 
alfa aesar™
H27111MD
250mg Each for $89.73
Description & Specifications

Specifications

Formula Weight 226.32
Chemical Name or Material 1-(1-Boc-3-azetidinyl)pyrrolidine
CAS 1019008-21-9
Assay 94%
Molecular Formula C12H22N2O2
MDL Number MFCD09860616
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C