Please login to your online account to display your discounted pricing

1,1-Bis(trichlorosilylmethyl)ethylene, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338650010

 View more versions of this product

Catalog No. AC338650010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Physical Form Liquid
Formula Weight 322.98
Quantity 1g
Color Clear colorless
TSCA TSCA
Chemical Name or Material 1, 1-Bis(trichlorosilylmethyl)ethylene
Molecular Formula C4 H6 Cl6 Si2
Linear Formula Cl3SiCH2C(CH2)CH2SiCl3
MDL Number MFCD00239510
CAS 78948-04-6
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4870 to 1.4890 (20°C, 589nm)
Synonym 2-Methylene-1, 3-propanediyl bis(trichlorosilane)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

Specifications

Chemical Name or Material 1, 1-Bis(trichlorosilylmethyl)ethylene
Assay ≥95.0%
Molecular Formula C4 H6 Cl6 Si2
Linear Formula Cl3SiCH2C(CH2)CH2SiCl3
Formula Weight 322.98
MDL Number MFCD00239510
Synonym 2-Methylene-1, 3-propanediyl bis(trichlorosilane)
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

SDS & Safety Documents