missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Bis(trichlorosilylmethyl)ethylene, ACROS Organics™

Chemical Identifiers

Molecular Formula C4 H6 Cl6 Si2
Linear Formula Cl3SiCH2C(CH2)CH2SiCl3
Formula Weight 322.98
MDL Number MFCD00239510
Synonym 2-Methylene-1, 3-propanediyl bis(trichlorosilane)
Chemical Name or Material 1, 1-Bis(trichlorosilylmethyl)ethylene
Assay ≥95.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC338650010 SDS Thermo Fisher Scientific
338650010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C4 H6 Cl6 Si2
322.98
2-Methylene-1, 3-propanediyl bis(trichlorosilane)
≥95.0%
Cl3SiCH2C(CH2)CH2SiCl3
MFCD00239510
1, 1-Bis(trichlorosilylmethyl)ethylene

Specifications

322.98
1g
C4 H6 Cl6 Si2
Cl3SiCH2C(CH2)CH2SiCl3
2-Methylene-1, 3-propanediyl bis(trichlorosilane)
Glass bottle
1, 1-Bis(trichlorosilylmethyl)ethylene
Liquid
Clear colorless
MFCD00239510
78948-04-6
1.4870 to 1.4890 (20°C, 589nm)
Authentic
≥95.0%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA