Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Bis(methylthio)-2-nitroethylene, 99%

$423.41 - $423.41

Chemical Identifiers

Chemical Name or Material 1,1-Bis (methylthio)-2-nitroethylene
CAS 13623-94-4
Molecular Formula C4H7NO2S2
Formula Weight 165.23
MDL Number MFCD00010443
Beilstein 1524538
Synonym 1,1-Bis(methylmercapto)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1178922

  SDS
alfa aesar™
A1178922
100g Each for $423.41
Description & Specifications

Specifications

Formula Weight 165.23
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,1-Bis (methylthio)-2-nitroethylene
CAS 13623-94-4
Synonym 1,1-Bis(methylmercapto)-2-nitroethylene
Beilstein 1524538
Molecular Formula C4H7NO2S2
MDL Number MFCD00010443
Melting Point 124° to 127°C
Odor Unpleasant
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.