missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Bis(hydroxymethyl)cyclopropane, TCI America™

$61.11 - $183.34

Chemical Identifiers

CAS 39590-81-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00464207
InChI Key YAINYZJQSQEGND-UHFFFAOYSA-N
Synonym 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
PubChem CID 4101437
IUPAC Name [1-(hydroxymethyl)cyclopropyl]methanol
SMILES C1CC1(CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B536525G View Documents TCI America
B536525G
25g Each for $183.34
 
B53655G View Documents TCI America
B53655G
5g Each for $61.11
 
Specifications

Chemical Identifiers

39590-81-3
102.133
YAINYZJQSQEGND-UHFFFAOYSA-N
4101437
C1CC1(CO)CO
C5H10O2
MFCD00464207
1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
[1-(hydroxymethyl)cyclopropyl]methanol

Specifications

98°C
25g
C5H10O2
1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
C1CC1(CO)CO
102.133
102.13
≥96.0% (GC)
Yellow
39590-81-3
MFCD00464207
YAINYZJQSQEGND-UHFFFAOYSA-N
[1-(hydroxymethyl)cyclopropyl]methanol
4101437
Clear Liquid at 20°C
1,1-Bis(hydroxymethyl)cyclopropane
Safety and Handling

Safety and Handling

TSCA : No