missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Bis(hydroxymethyl)cyclopropane, 90%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC426250010

 View more versions of this product

Catalog No. AC426250010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

102.13
1.065g/mL
100.0
1, 1-Bis(hydroxymethyl)cyclopropane
39590-81-3
Glass bottle
1g
88.0
90%
C5H10O2
MFCD00464207

Chemical Identifiers

39590-81-3
100.0
C5H10O2
MFCD00464207
88.0
90%
102.13
1, 1-Bis(hydroxymethyl)cyclopropane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.