Please login to your online account to display your discounted pricing

1,1-Bis(hydroxymethyl)cyclopropane, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 1-Bis(hydroxymethyl)cyclopropane
CAS 39590-81-3
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C5H10O2
Formula Weight 102.13
MDL Number MFCD00464207
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426250010

  SDS
thermo fisher scientific
AC426250010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426250050

  SDS
thermo fisher scientific
AC426250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.13
Quantity 1g
Density 1.065g/mL
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Chemical Name or Material 1, 1-Bis(hydroxymethyl)cyclopropane
Molecular Formula C5H10O2
CAS 39590-81-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00464207
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.