missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten, 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4428803

 View more versions of this product

Catalog No. AA4428803



Specifications

Specifications

[1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonyltungsten
220°C (decomposition)
67292-26-6
MFCD03427160
850.28
98%
Air sensitive
1g
C38H28FeO4P2W
UN3466
Powder

Chemical Identifiers

67292-26-6
C38H28FeO4P2W
MFCD03427160
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents