Please login to your online account to display your discounted pricing

Alfa Aesar™ [1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten, 98%

Chemical Identifiers

Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonyltungsten
CAS 67292-26-6
Molecular Formula C38H28FeO4P2W
Formula Weight 850.28
MDL Number MFCD03427160
UN Number UN3466
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4428803

  SDS
alfa aesar™
4428803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4428806

  SDS
alfa aesar™
4428806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 850.28
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonyltungsten
Assay 98%
Sensitivity Air Sensitive
Melting Point 220°C (dec.)
Health Hazard 2 GHS H Statement
H301-H331-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
CAS 67292-26-6
UN Number UN3466
Physical Form Powder
MDL Number MFCD03427160
Molecular Formula C38H28FeO4P2W
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.