Please login to your online account to display your discounted pricing

Alfa Aesar™ [1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonylmolybdenum, 98%

$45.20 - $197.39

Chemical Identifiers

Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylmolybdenum
CAS 67292-28-8
Molecular Formula C38H28FeMoO4P2
Formula Weight 762.37
MDL Number MFCD03427159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4428103

  SDS
alfa aesar™
4428103
1g Each for $45.20

AA4428106

  SDS
alfa aesar™
4428106
5g Each for $197.39
Description & Specifications

Specifications

Formula Weight 762.37
Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylmolybdenum
Physical Form Powder
Assay 98%
Molecular Formula C38H28FeMoO4P2
MDL Number MFCD03427159
CAS 67292-28-8
Melting Point 256°C (dec.)
Quantity 1g