Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1-Bis(diphenylphosphino)ethylene, 98%

Chemical Identifiers

Chemical Name or Material 1,1-Bis(diphenylphosphino)ethylene
CAS 84494-89-3
Linear Formula [(C6H5)2P]2C=CH2
Formula Weight 396.41g/mol
MDL Number MFCD00191809
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5586403

 
alfa aesar™
H55864-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5586406

 
alfa aesar™
H55864-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 110 to 111°C
Quantity 1g
Chemical Name or Material 1,1-Bis(diphenylphosphino)ethylene
CAS 84494-89-3
Physical Form Solid
Assay 98%
Linear Formula [(C6H5)2P]2C=CH2
Formula Weight 396.41g/mol
MDL Number MFCD00191809