Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Bis(diisopropylphosphino)ferrocene, 98%

$206.34 - $593.69

Chemical Identifiers

Chemical Name or Material 1,1'-Bis(diisopropylphosphino)ferrocene
CAS 97239-80-0
Molecular Formula C22H36FeP2
Formula Weight 418.34g/mol
MDL Number MFCD02684559
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705706

 
alfa aesar™
H27057-06
5g Each for $593.69

AAH2705703

 
alfa aesar™
H27057-03
1g Each for $206.34
Description & Specifications

Specifications

Melting Point 50° to 52°C
Quantity 5g
Flash Point >110°C (230°F)
Chemical Name or Material 1,1'-Bis(diisopropylphosphino)ferrocene
CAS 97239-80-0
Assay 98%
Molecular Formula C22H36FeP2
Formula Weight 418.34g/mol
MDL Number MFCD02684559

Applications
The general and efficient cross-coupling of thiols with aryl halides was developed utilizing Pd(OAc) 2 /1,1′-bis(diisopropylphosphino)ferrocene as the catalyst. The oxidative electrochemistry of dippf was examined, and it was found to follow an ECDim mechanism. Seven new compounds containing dippf were prepared and characterized spectroscopically. The crystal structures of [MCl2(dippf)] (M = Pt and Zn) were determined.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.