Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Bis(dichlorophosphino)ferrocene, 98%

Chemical Identifiers

Chemical Name or Material 1,1'-Bis(dichlorophosphino)ferrocene
CAS 142691-70-1
Molecular Formula C10H8Cl4FeP2
Formula Weight 387.78g/mol
MDL Number MFCD01630854
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2729303

 
alfa aesar™
H27293-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 1,1'-Bis(dichlorophosphino)ferrocene
CAS 142691-70-1
Assay 98%
Molecular Formula C10H8Cl4FeP2
Formula Weight 387.78g/mol
MDL Number MFCD01630854
UN Number UN3261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.