Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1'-Bis(di-tert-butylphosphino)ferrocene, 98%

$69.82 - $238.91

Chemical Identifiers

Chemical Name or Material 1,1'-Bis(di-tert-butylphosphino)ferrocene
CAS 84680-95-5
Molecular Formula C26H44FeP2
Formula Weight 474.43g/mol
MDL Number MFCD01630818
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1975904

 
alfa aesar™
L19759-04
2g Each for $238.91

AAL19759ME

 
alfa aesar™
L19759-ME
500mg Each for $69.82
Description & Specifications

Specifications

Melting Point 73° to 78°C
Quantity 2g
Chemical Name or Material 1,1'-Bis(di-tert-butylphosphino)ferrocene
CAS 84680-95-5
Assay 98%
Molecular Formula C26H44FeP2
Formula Weight 474.43g/mol
MDL Number MFCD01630818
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
The rate of palladium-catalyzed amination of unactivated aryl chlorides is accelerated by sterically hindered chelating alkyl phosphines, ie, 1,1′-bis(di-tert-butylphosphino)ferrocene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.