missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[1,1«-Biphenyl]-3-ylboronic acid, 97%

Chemical Identifiers

CAS 5122-95-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C12H11BO2
Formula Weight 198.03
Chemical Name or Material Biphenyl-3-boronic acid
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC358880010 SDS Thermo Fisher Scientific
AC358880010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC358882500 SDS Thermo Fisher Scientific
35888-2500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5122-95-2
100.0
C12H11BO2
Biphenyl-3-boronic acid
97.5
98%
198.03
≥97.5%

Specifications

Powder
1g
100.0
97.5
Authentic
5122-95-2
≥97.5%
198.03
Light beige
C12H11BO2
Glass bottle
98%
Biphenyl-3-boronic acid

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.