Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1,1'-Bi(2-naphthyl) dimethanesulfonate, 97%

$468.88 - $468.88

Chemical Identifiers

Chemical Name or Material (+/-)-1,1'-Bi(2-naphthyl) dimethanesulfonate
CAS 182568-57-6
Molecular Formula C22H18O6S2
Formula Weight 442.50g/mol
MDL Number MFCD05865182
Synonym 1,1'-Binaphthalene-2,2'-diol dimethanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5108706

 
alfa aesar™
H51087-06
Each for $468.88
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material (+/-)-1,1'-Bi(2-naphthyl) dimethanesulfonate
CAS 182568-57-6
Assay 97%
Molecular Formula C22H18O6S2
Formula Weight 442.50g/mol
MDL Number MFCD05865182
Synonym 1,1'-Binaphthalene-2,2'-diol dimethanesulfonate