Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1,1'-Bi(2-naphthol), 99%

$455.04 - $455.04

Chemical Identifiers

Chemical Name or Material (±)-1,1'-Bi (2-naphthol)
CAS 602-09-5
Molecular Formula C20H14O2
Formula Weight 286.33
MDL Number MFCD00004068
UN Number UN2811
Beilstein 997518
Merck Index 14,1226
Synonym (+/-)-Binaphthalene-2,2'-diol; (+/-)-BINOL
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1118222

  SDS
alfa aesar™
A1118222
100g Each for $455.04

AAA1118206

  SDS
alfa aesar™
A1118206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1118214

  SDS
alfa aesar™
A1118214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.33
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material (±)-1,1'-Bi (2-naphthol)
TSCA TSCA
Assay 99%
Molecular Formula C20H14O2
MDL Number MFCD00004068
CAS 602-09-5
UN Number UN2811
Beilstein 997518
Melting Point 215° to 218°C
Merck Index 14,1226
Synonym (+/-)-Binaphthalene-2,2'-diol; (+/-)-BINOL
Quantity 100g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Odor Characteristic
RTECS Number DU3106000

Applications
(+/-)-1,1′-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.