Please login to your online account to display your discounted pricing

1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC24805DA

 View more versions of this product

Catalog No. CC24805DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 255.13
Quantity 1g
Assay 97%
Molecular Formula C10H7BrOS
Packaging Amber Glass Bottle
Chemical Name or Material 1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone
CAS 1131-87-9
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7BrOS
Formula Weight 255.13