Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride, 97%

Chemical Identifiers

Chemical Name or Material 1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride
CAS 1134322-93-2
Formula Weight 411.34
MDL Number MFCD03844681
Synonym 1-[1-[(4-Fluorophenyl)methyl]-1H-benzimidazol-2-yl]-N-methyl-4-piperidinamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63927MD

 
alfa aesar™
H63927MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 411.34
Chemical Name or Material 1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride
Assay 97%
Quantity 250mg
Storage Note 1 2° to 8°C
CAS 1134322-93-2
Physical Form Solid
Synonym 1-[1-[(4-Fluorophenyl)methyl]-1H-benzimidazol-2-yl]-N-methyl-4-piperidinamine dihydrochloride
MDL Number MFCD03844681
Safety and Handling

2° to 8°C