Learn More
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1543336
Description
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes
Solubility
Miscible in water.
Notes
Air & Moisture Sensitive. Store under inert gas. Incompatible with strong oxidizing agents, strong acids.
Specifications
1,1,4,7,7-Pentamethyldiethylenetriamine | |
<-20°C | |
199°C | |
C9H23N3 | |
MFCD00014876 | |
UN2922 | |
Air and moisture sensitive | |
Miscible in water. | |
CN(C)CCN(C)CCN(C)C | |
173.304 | |
CHEBI:39475 | |
98% |
3030-47-5 | |
0.83 | |
72°C (161°F) | |
1.442 | |
500 g | |
1741396 | |
pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine | |
UKODFQOELJFMII-UHFFFAOYSA-N | |
N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine | |
18196 | |
173.3 |
Chemical Identifiers
3030-47-5 | |
173.304 | |
UKODFQOELJFMII-UHFFFAOYSA-N | |
18196 | |
N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine |
C9H23N3 | |
MFCD00014876 | |
pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine | |
CHEBI:39475 | |
CN(C)CCN(C)CCN(C)C |
Safety and Handling
GHS H Statement
H311-H314-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Combustible liquid.
Harmful if swallowed.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H311-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 221-201-1
RTECSNumber : IE2100000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only