missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,4,7,10,10-Hexamethyltriethylenetetramine 98.0+%, TCI America™
Supplier: TCI America H137125ML
Specifications
| 1,1,4,7,10,10-Hexamethyltriethylenetetramine | |
| Yellow | |
| C12H30N4 | |
| 25 mL | |
| 1,1,4,7,10,10-hexamethyltriethylenetetramine, hexamethyltriethylenetetramine, 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane, hmteta, 1,2-ethanediamine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethyl, n1,n1'-ethane-1,2-diyl bis n1,n2,n2-trimethylethane-1,2-diamine, n,n,n',n,n',n'-hexamethyltriethylenetetramine, triethylenetetramine, 1,1,4,7,10,10-hexamethyl, 1,1,4,7,10,10-hexamethyl-triethylenetetramine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethylethylenediamine | |
| CN(C)CCN(C)CCN(C)CCN(C)C | |
| 230.40 | |
| 230.40 | |
| Liquid |
| 3083-10-1 | |
| 130°C | |
| MFCD00025684 | |
| 2735 | |
| DWFKOMDBEKIATP-UHFFFAOYSA-N | |
| N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine | |
| 76509 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3083-10-1 | |
| 230.40 | |
| DWFKOMDBEKIATP-UHFFFAOYSA-N | |
| 76509 | |
| CN(C)CCN(C)CCN(C)CCN(C)C |
| C12H30N4 | |
| MFCD00025684 | |
| 1,1,4,7,10,10-hexamethyltriethylenetetramine, hexamethyltriethylenetetramine, 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane, hmteta, 1,2-ethanediamine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethyl, n1,n1'-ethane-1,2-diyl bis n1,n2,n2-trimethylethane-1,2-diamine, n,n,n',n,n',n'-hexamethyltriethylenetetramine, triethylenetetramine, 1,1,4,7,10,10-hexamethyl, 1,1,4,7,10,10-hexamethyl-triethylenetetramine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethylethylenediamine | |
| N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine |
Safety and Handling
EINECSNumber : (7)-0005
RTECSNumber : KH8587135
TSCA : Yes