missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%

Manufacturer:  Alfa Aesar™ H5608914

 View more versions of this product

Catalog No. AAH5608914



Description

Description

Applications
1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

1,1,4,7,10,10-Hexamethyltriethylenetetramine
130°C (11mmHg)
3083-10-1
MFCD00025684
1,1,4,7,10,10-hexamethyl-triethylenetetramine
CN(C)CCN(C)CCN(C)CCN(C)C
230.40
230.39
102°C (216°F)
25g
C12H30N4
UN3725
DWFKOMDBEKIATP-UHFFFAOYSA-N
N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine
76509
97%

Chemical Identifiers

3083-10-1
230.40
DWFKOMDBEKIATP-UHFFFAOYSA-N
76509
CN(C)CCN(C)CCN(C)CCN(C)C
C12H30N4
MFCD00025684
1,1,4,7,10,10-hexamethyl-triethylenetetramine
N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : KH8587135

TSCA : TSCA

Documents