Please login to your online account to display your discounted pricing

1,1'-(4,6-Dihydroxy-1,3-phenylene)bisethanone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1,1'-(4,6-Dihydroxy-1,3-phenylene)bisethanone
CAS 2161-85-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H10O4
Linear Formula (HO)C6H(COCH3)
Formula Weight 194.19
MDL Number MFCD00017655
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372950050

  SDS
thermo fisher scientific
AC372950050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 194.19
Quantity 5g
Color White to off-white
Melting Point 178° to 185°C
CAS Max % 100.0
Synonym 4, 6-Diacetylresorcinol
Linear Formula (HO)C6H(COCH3)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00017655
Chemical Name or Material 1,1'-(4,6-Dihydroxy-1,3-phenylene)bisethanone
Molecular Formula C10H10O4
CAS 2161-85-5
Assay Percent Range 99%
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Infrared Spectrum Authentic