Learn More
1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, Thermo Scientific Chemicals
$81.30 - $204.53
Chemical Identifiers
CAS | 1450-63-1 |
---|---|
Molecular Formula | C28H22 |
Molecular Weight (g/mol) | 358.48 |
MDL Number | MFCD00004766 |
InChI Key | KLCLIOISYBHYDZ-UHFFFAOYSA-N |
Synonym | 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene |
PubChem CID | 74060 |
SMILES | C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1722503
|
Thermo Scientific Chemicals
A1722503 |
1 g |
N/A
|
|
|||||
AAA1722506
|
Thermo Scientific Chemicals
A1722506 |
5 g |
N/A
|
|
|||||
Description
1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.
Solubility
Soluble in ethanol, benzene, chloroform, tolune and acetic acid.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
1450-63-1 | |
358.48 | |
KLCLIOISYBHYDZ-UHFFFAOYSA-N | |
74060 |
C28H22 | |
MFCD00004766 | |
1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene | |
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
1450-63-1 | |
C28H22 | |
1 g | |
Soluble in ethanol,benzene,chloroform,tolune and acetic acid. | |
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 | |
358.48 | |
358.48 | |
1,1,4,4-Tetraphenyl-1,3-butadiene |
197°C to 202°C | |
MFCD00004766 | |
1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene | |
KLCLIOISYBHYDZ-UHFFFAOYSA-N | |
1,4,4-triphenylbuta-1,3-dienylbenzene | |
74060 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-914-7
RTECSNumber : CY9040630
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only