Please login to your online account to display your discounted pricing

1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthaline, Technical Grade, Maybridge

Chemical Identifiers

Chemical Name or Material 1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene
Assay 90%
Assay Percent Range 90%
Molecular Formula C14H20
Formula Weight 188.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00201EE

 
fisher scientific company
TL00201EE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.31
Quantity 50g
Assay 90%
Chemical Name or Material 1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene
Molecular Formula C14H20
CAS 6683-46-1
Assay Percent Range 90%