Please login to your online account to display your discounted pricing

1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthaline, Technical Grade, Maybridge


Quantity 50g

Manufacturer: fisher scientific company  TL00201EE

 View more versions of this product

Catalog No. TL00201EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 188.31
Quantity 50g
Assay 90%
CAS 6683-46-1
Chemical Name or Material 1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene
Molecular Formula C14H20
Assay Percent Range 90%

Specifications

Chemical Name or Material 1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene
Assay 90%
Assay Percent Range 90%
Molecular Formula C14H20
Formula Weight 188.31