missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,3-Triphenylpropargyl alcohol, 97%

Manufacturer:  Alfa Aesar™ H5346106

 View more versions of this product

Catalog No. AAH5346106



Specifications

Specifications

1,1,3-Triphenylpropargyl alcohol
5g
C21H16O
1,1,3-triphenyl-2-propyn-1-ol
C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)(C3=CC=CC=C3)O
284.358
284.36
80°C to 82°C
1522-13-0
MFCD00004449
VWRQCJRTHKUVNF-UHFFFAOYSA-N
1,1,3-triphenylprop-2-yn-1-ol
137058
97%

Chemical Identifiers

1522-13-0
284.358
VWRQCJRTHKUVNF-UHFFFAOYSA-N
137058
C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)(C3=CC=CC=C3)O
C21H16O
MFCD00004449
1,1,3-triphenyl-2-propyn-1-ol
1,1,3-triphenylprop-2-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents