missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,3-Triphenyl-2-propyn-1-ol, TCI America™

Manufacturer:  TCI America T241825G

 View more versions of this product

Catalog No. T241825G



Specifications

Specifications

1,1,3-Triphenyl-2-propyn-1-ol
White-Yellow
25g
C21H16O
1,1,3-triphenylpropargyl alcohol, 1,1,3-triphenyl-2-propyn-1-ol, diphenyl phenylethynyl carbinol, benzenemethanol, .alpha.-phenyl-.alpha.-phenylethynyl, triphenylprop-2-yn-1-ol, 1,1,3-triphenylpropargylalcohol, 2-propyn-1-ol, 1,1,3-triphenyl, pubchem14025, acmc-209d7i, triphenyl-1-propyne-3-ol
C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)(C3=CC=CC=C3)O
284.358
284.36
≥98.0% (GC)
220°C
82°C
1522-13-0
MFCD00004449
VWRQCJRTHKUVNF-UHFFFAOYSA-N
1,1,3-triphenylprop-2-yn-1-ol
137058
Crystal-Powder at 20°C

Chemical Identifiers

1522-13-0
284.358
VWRQCJRTHKUVNF-UHFFFAOYSA-N
137058
C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)(C3=CC=CC=C3)O
C21H16O
MFCD00004449
1,1,3-triphenylpropargyl alcohol, 1,1,3-triphenyl-2-propyn-1-ol, diphenyl phenylethynyl carbinol, benzenemethanol, .alpha.-phenyl-.alpha.-phenylethynyl, triphenylprop-2-yn-1-ol, 1,1,3-triphenylpropargylalcohol, 2-propyn-1-ol, 1,1,3-triphenyl, pubchem14025, acmc-209d7i, triphenyl-1-propyne-3-ol
1,1,3-triphenylprop-2-yn-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents