Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,3-Triethoxypropane, 98+%

Chemical Identifiers

Chemical Name or Material 1,1,3-Triethoxypropane
CAS 7789-92-6
Molecular Formula CH3CH2O(CH2)2CH(OCH2CH3)2
Formula Weight 176.26
MDL Number MFCD00009241
UN Number UN3271
Beilstein 1098506
Synonym 3-Ethoxypropanal diethyl acetal; 3-Ethoxypropionaldehyde diethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1301106

 
alfa aesar™
L1301106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1301114

 
alfa aesar™
L1301114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Chemical Name or Material 1,1,3-Triethoxypropane
Molecular Formula CH3CH2O(CH2)2CH(OCH2CH3)2
MDL Number MFCD00009241
Beilstein 1098506
Synonym 3-Ethoxypropanal diethyl acetal; 3-Ethoxypropionaldehyde diethyl acetal
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Flash Point 53°C (127°F)
RTECS Number TZ9450000
Quantity 5g
Assay ≥98%
CAS 7789-92-6
UN Number UN3271
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 184° to 186°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.