missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,3-Triethoxypropane, 98+%

Manufacturer:  Alfa Aesar™ L1301106

 View more versions of this product

Catalog No. AAL1301106



Specifications

Specifications

1,1,3-Triethoxypropane
184°C to 186°C
7789-92-6
MFCD00009241
1098506
LGICWIVABSMSDK-UHFFFAOYSA-N
1,1,3-triethoxypropane
24624
≥98%
53°C (127°F)
5g
C9H20O3
UN3271
3-ethoxypropionaldehyde diethyl acetal, propane, 1,1,3-triethoxy, 1,3,3-triethoxypropane, 3-ethoxypropanal diethyl acetal, propane, 1,3,3-triethoxy, unii-1qw8j6vxam, 3-ethoxypropionaldehydediethylacetal, 1qw8j6vxam, beta-ethoxypropionaldehyde diethyl acetal, .beta.-ethoxypropionaldehyde diethyl acetal
CCOCCC(OCC)OCC
176.256
176.26

Chemical Identifiers

7789-92-6
176.256
LGICWIVABSMSDK-UHFFFAOYSA-N
24624
CCOCCC(OCC)OCC
C9H20O3
MFCD00009241
3-ethoxypropionaldehyde diethyl acetal, propane, 1,1,3-triethoxy, 1,3,3-triethoxypropane, 3-ethoxypropanal diethyl acetal, propane, 1,3,3-triethoxy, unii-1qw8j6vxam, 3-ethoxypropionaldehydediethylacetal, 1qw8j6vxam, beta-ethoxypropionaldehyde diethyl acetal, .beta.-ethoxypropionaldehyde diethyl acetal
1,1,3-triethoxypropane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

RTECSNumber : TZ9450000

Documents