missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-((1,3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™
Chemical Identifiers
CAS | 864754-17-6 |
---|---|
Molecular Formula | C13H21BN2O4 |
Molecular Weight (g/mol) | 280.13 |
InChI Key | IWEGDQUCWQFKHS-UHFFFAOYSA-N |
Synonym | 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-1,3 dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-yl methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3-dioxolan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pubchem18608 |
PubChem CID | 17998927 |
IUPAC Name | 1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3OCCO3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC444990010
|
Thermo Fisher Scientific
AC444990010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC444990050
|
Thermo Fisher Scientific
AC444990050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
864754-17-6 | |
280.13 | |
1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-1,3 dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-yl methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3-dioxolan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pubchem18608 | |
1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
C13H21BN2O4 | |
IWEGDQUCWQFKHS-UHFFFAOYSA-N | |
17998927 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3OCCO3 |
Specifications
Glass Bottle | |
1g | |
96% min. (GC) | |
1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-1,3 dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-yl methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3-dioxolan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pubchem18608 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3OCCO3 | |
280.13 | |
280.13 | |
1-((1, 3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester |
63°C to 64°C | |
864754-17-6 | |
C13H21BN2O4 | |
IWEGDQUCWQFKHS-UHFFFAOYSA-N | |
1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole | |
17998927 | |
97% |
Safety and Handling
GHS Signal Word: Warning