missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-((1,3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444990050

 View more versions of this product

Catalog No. AC444990050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-((1, 3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester
5g
96% min. (GC)
1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-1,3 dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-yl methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3-dioxolan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pubchem18608
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3OCCO3
280.13
280.13
63°C to 64°C
864754-17-6
C13H21BN2O4
IWEGDQUCWQFKHS-UHFFFAOYSA-N
1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
17998927
97%

Chemical Identifiers

864754-17-6
280.13
1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-1,3 dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-yl methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1,3-dioxolan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-1,3-dioxolan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pubchem18608
1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
C13H21BN2O4
IWEGDQUCWQFKHS-UHFFFAOYSA-N
17998927
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3OCCO3
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents