missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H7BrO3
Formula Weight 243.06
Chemical Name or Material 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC01605EA Fisher Scientific Company
CC01605EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01605DA SDS Fisher Scientific Company
CC01605DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01605CB SDS Fisher Scientific Company
CC01605CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
243.06
97%
C9H7BrO3
1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one

Specifications

40288-65-1
97%
243.06
1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one
Amber Glass Bottle
C9H7BrO3
10g
97%