Please login to your online account to display your discounted pricing

1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC01605DA

 View more versions of this product

Catalog No. CC01605DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 243.06
Quantity 1g
Assay 97%
Chemical Name or Material 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H7BrO3
CAS 40288-65-1

Specifications

Chemical Name or Material 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7BrO3
Formula Weight 243.06