Please login to your online account to display your discounted pricing

1,1,3,3-Tetramethylguanidine, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  221290050

 View more versions of this product

Catalog No. AC221290050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Molecular Formula C5H13N3
Linear Formula (CH3)2NC(=NH)N(CH3)2
Formula Weight 115.18
CAS 80-70-6
Density 0.9100g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4670 to 1.4690 (20°C, 589nm)
Water 0.5% (K.F.) max.
Packaging Glass bottle
Fieser 01,1145; 04,489; 12,477

Specifications

Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H13N3
Linear Formula (CH3)2NC(=NH)N(CH3)2
Formula Weight 115.18