missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,3,3-Tetramethylguanidine, 99%, ACROS Organics™

$96.33 - $324.67

Chemical Identifiers

CAS 80-70-6
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
InChI Key KYVBNYUBXIEUFW-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyl guanidine, 1,1,3,3-tetramethyl-guanidine, guanidine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-guanidine, n,n,n',n'-tetramethylguanidine, n,n-1,1,3,3-tetramethylguanidine, tetramethyl guanidine, tetramethylguanidine, unii-vez101e7zu, vez101e7zu
PubChem CID 66460
IUPAC Name 1,1,3,3-tetramethylguanidine
SMILES CN(C)C(=N)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221291000
SDS View Documents
Thermo Fisher Scientific
221291000
100g Glass bottle Each for $96.33
 
AC221295000
SDS View Documents
Thermo Fisher Scientific
221295000
500g Glass bottle Each for $324.67
 
Specifications

Chemical Identifiers

80-70-6
115.18
1,1,3,3-tetramethyl guanidine, 1,1,3,3-tetramethyl-guanidine, guanidine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-guanidine, n,n,n',n'-tetramethylguanidine, n,n-1,1,3,3-tetramethylguanidine, tetramethyl guanidine, tetramethylguanidine, unii-vez101e7zu, vez101e7zu
1,1,3,3-tetramethylguanidine
C5H13N3
KYVBNYUBXIEUFW-UHFFFAOYSA-N
66460
CN(C)C(=N)N(C)C

Specifications

0.9100g/mL
Authentic
1.4670 to 1.4690
0.002 Pas (20°C)
160.0°C to 162.0°C
-30.0°C
80-70-6
C5H13N3
04,I,335
15,9375
Solubility in water: miscible. Other solubilities: miscible
CN(C)C(=N)N(C)C
115.18
115.18
99%
50°C
Glass bottle
0.91
0.5% Max. (K.F.)
Colorless to Yellow
100g
98.5% min. (GC)
(CH3)2NC(=NH)N(CH3)2
01,1145; 04,489; 12,477
1,1,3,3-tetramethyl guanidine, 1,1,3,3-tetramethyl-guanidine, guanidine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-guanidine, n,n,n',n'-tetramethylguanidine, n,n-1,1,3,3-tetramethylguanidine, tetramethyl guanidine, tetramethylguanidine, unii-vez101e7zu, vez101e7zu
KYVBNYUBXIEUFW-UHFFFAOYSA-N
1,1,3,3-tetramethylguanidine
66460
Liquid
1,1,3,3-Tetramethylguanidine
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov

GHS Signal Word: Danger

EINECSNumber : 201-302-7