Please login to your online account to display your discounted pricing

1,1,3,3-Tetramethylguanidine, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  221291000

 View more versions of this product

Catalog No. AC221291000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Physical Form Liquid
Formula Weight 115.18
Quantity 100g
Color Clear colorless to yellow
Water 0.5% Max. (K.F.)
Density 0.9100g/mL
Fieser 01,1145; 04,489; 12,477
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C5H13N3
Linear Formula (CH3)2NC(=NH)N(CH3)2
Refractive Index 1.4670 to 1.4690 (20°C, 589nm)
CAS 80-70-6
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Assay Percent Range 99%

Specifications

Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H13N3
Linear Formula (CH3)2NC(=NH)N(CH3)2
Formula Weight 115.18