Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,3,3-Tetramethyl-1,3-divinyldisilazane, 96%

Chemical Identifiers

Chemical Name or Material 1,1,3,3-Tetramethyl-1,3-divinyldisilazane
CAS 7691-02-3
Molecular Formula C8H19NSi2
Formula Weight 185.42
MDL Number MFCD00042846
UN Number UN2924
Beilstein 2411661
Synonym 1,3-Divinyl-1,1,3,3-tetramethyldisilazane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1648006

 
alfa aesar™
L1648006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1648014

 
alfa aesar™
L1648014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1648022

 
alfa aesar™
L1648022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.42
Molecular Formula C8H19NSi2
CAS 7691-02-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Chemical Name or Material 1,1,3,3-Tetramethyl-1,3-divinyldisilazane
TSCA TSCA
Assay 96%
MDL Number MFCD00042846
UN Number UN2924
Beilstein 2411661
Synonym 1,3-Divinyl-1,1,3,3-tetramethyldisilazane
Flash Point 42°C (107°F)
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
RTECS Number VV1302250
Boiling Point 160° to 161°C
Quantity 5g

Applications
1,1,3,3-Tetramethyl-1,3-divinyldisilazane is used for silicone rubber, silicone resin, silicone gel and vinyl manufacturing.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.