Please login to your online account to display your discounted pricing

1,1,3,3,5,5-Hexamethyltrisiloxane, 97%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  337570050

 View more versions of this product

Catalog No. AC337570050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 208.48
Quantity 5mL
Color Clear colorless
Density 0.8220g/mL
EINECS Number 214-716-8
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
Name Note 97%
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H20O2Si3
Refractive Index 1.3790 to 1.3820 (20°C, 589nm)
Linear Formula Si(CH3)2[OSi(CH3)2H]
MDL Number MFCD00039788
CAS 1189-93-1
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
Name Note 97%
CAS 1189-93-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H20O2Si3
Linear Formula Si(CH3)2[OSi(CH3)2H]
Formula Weight 208.48
MDL Number MFCD00039788
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents