Please login to your online account to display your discounted pricing

1,1,3,3,5,5-Hexamethyltrisiloxane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
Name Note 97%
CAS 1189-93-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H20O2Si3
Linear Formula Si(CH3)2[OSi(CH3)2H]
Formula Weight 208.48
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC337570050

  SDS
thermo fisher scientific
337570050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 208.48
Quantity 5mL
Color Clear colorless
TSCA TSCA
EINECS Number 214-716-8
Density 0.8220g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.3790 to 1.3820 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Name Note 97%
Molecular Formula C6H20O2Si3
CAS 1189-93-1
Linear Formula Si(CH3)2[OSi(CH3)2H]
Assay Percent Range 97%
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
MDL Number MFCD00039788

Reduction reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.