Learn More
1,1,2-Trimethyl-1H-benzo[e]indole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2180406
Description
Reactant used for preparation of fluorescent probes for imaging of tumors, hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant used for preparation of fluorescent probes for imaging of tumors , hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
1,1,2-Trimethyl-1H-benzo[e]indole | |
114°C to 119°C | |
MFCD00082627 | |
153709 | |
Slightly soluble in water. | |
CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C | |
209.29 | |
209.29 |
41532-84-7 | |
C15H15N | |
5 g | |
1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole | |
WJZSZXCWMATYFX-UHFFFAOYSA-N | |
1,1,2-trimethylbenzo[e]indole | |
170530 | |
97% |
Chemical Identifiers
41532-84-7 | |
209.29 | |
WJZSZXCWMATYFX-UHFFFAOYSA-N | |
170530 | |
CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C |
C15H15N | |
MFCD00082627 | |
1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole | |
1,1,2-trimethylbenzo[e]indole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 255-429-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only